missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,4-difluorobenzoic acid, 97%, Thermo Scientific™

Catalog Number 11441137
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
154257-75-7
C7H2ClF2O2
191.54
MFCD01631387
YGYZTZAEZMCPSC-UHFFFAOYSA-M
3-chloro-2,4-difluoro-benzoic acid, 2,4-difluoro-3-chlorobenzoic acid, 3-chloro-2,4-difluorobenzoicacid, benzoic acid, 3-chloro-2,4-difluoro, pubchem2217, acmc-1byme, intermediates-zcf02173, ksc181k3p
2773532
3-chloro-2,4-difluorobenzoic acid
[O-]C(=O)C1=CC=C(F)C(Cl)=C1F
This item is not returnable. View return policy
154257-75-7
C7H2ClF2O2
191.54
MFCD01631387
YGYZTZAEZMCPSC-UHFFFAOYSA-M
3-chloro-2,4-difluoro-benzoic acid, 2,4-difluoro-3-chlorobenzoic acid, 3-chloro-2,4-difluorobenzoicacid, benzoic acid, 3-chloro-2,4-difluoro, pubchem2217, acmc-1byme, intermediates-zcf02173, ksc181k3p
2773532
3-chloro-2,4-difluorobenzoic acid
[O-]C(=O)C1=CC=C(F)C(Cl)=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 174°C to 177°C
Quantity 1 g
Beilstein 7290655
Formula Weight 192.55
Percent Purity 97%
Chemical Name or Material 3-Chloro-2,4-difluorobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'