missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1H-indazole, 99%

Product Code. 10073370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10073370 25 g Glass bottle 25g
10267020 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10073370 Supplier Thermo Scientific Acros Supplier No. 263740250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 29110-74-5 | C7H5ClN2 | 152.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29110-74-5
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD00067527
InChI Key QPHAGNNWDZSKJH-UHFFFAOYSA-N
Synonym 3-chloro-1h-indazole, 3-chloroindazole, 1h-indazole, 3-chloro, 2h-indazole, 3-chloro, 1h-indazole,3-chloro, acmc-1cio6, ksc205k5h
PubChem CID 95894
IUPAC Name 3-chloro-2H-indazole
SMILES ClC1=C2C=CC=CC2=NN1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 148°C to 150°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Formula Weight 152.58
Percent Purity 99%
Physical Form Fluffy Powder
Chemical Name or Material 3-Chloro-1H-indazole, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chloro-1H-indazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.