missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1,2-propanediol, TRC

Product Code. 18073420
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
Unit Size:
100mg
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18073420 1 g 1g
18020420 25 mg 25mg
18021910 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18073420 Supplier Toronto Research Chemicals Supplier No. TRCC3797101G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-24-2
Molecular Formula C3 H7 Cl O2
Molecular Weight (g/mol) 110.5395
Synonym 3-Chloro-1,2-propanediol, (RS)-3-Chloro-1,2-propanediol, (RS)-α-Chlorohydrin, (±)-2,3-Dihydroxychloropropane, (±)-3-Chloro-1,2-propanediol, 1,2-Dihydroxy-3-chloropropane, 1-Chloro-1-deoxyglycerol, 1-Chloro-2,3-dihydroxypropane, 1-Chloro-2,3-propanediol, 3-Chloro-1,2-dihydroxypropane, 3-Chloro-1,2-propylene glycol, 3-Chloropropanediol, 3-Chloropropylene glycol, 3-Monochloro-1,2-propanediol, Chlorohydrin, Chloropropanediol, Ekorod A, Epibloc, Glycerin epichlorohydrin, Glycerin monochlorohydrin, Glycerin α-monochlorhydrin, Glycerol 3-chlorohydrin, Glycerol monochlorohydrin, Glycerol α-chlorohydrin, Glycerol α-monochlorohydrin, Glyceryl chloride, Glyceryl α-chlorohydrin, U 5897, dl-α-Chlorohydrin, α-Chlorohydrin, α-Monochlorohydrin
IUPAC Name 3-chloropropane-1,2-diol
SMILES OCC(O)CCl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C3H7ClO2/c4-1-3(6)2-5/h3,5-6H,1-2H2
Formula Weight 110.0135
Percent Purity >95%
Chemical Name or Material 3-Chloropropane-1,2-diol
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.