missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1,2-propanediol, 98%

Product Code. 10163670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
25mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10163670 500 mL Glass bottle 500mL
10591961 25 mL Glass bottle 25mL
10295190 1 L Glass bottle 1 L
10658283 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10163670 Supplier Thermo Scientific Acros Supplier No. 148125000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-24-2 | C3H7ClO2 | 110.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-24-2
Molecular Formula C3H7ClO2
Molecular Weight (g/mol) 110.54
MDL Number MFCD00004712
InChI Key SSZWWUDQMAHNAQ-UHFFFAOYNA-N
Synonym 3-chloro-1,2-propanediol, alpha-chlorohydrin, chlorodeoxyglycerol, glycerol alpha-monochlorohydrin, epibloc, 1,2-propanediol, 3-chloro, a-chlorohydrin, 3-chloropropanediol, 3-chloropropylene glycol, glycerol chlorohydrin
PubChem CID 7290
ChEBI CHEBI:18721
IUPAC Name 3-chloropropane-1,2-diol
SMILES OCC(O)CCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -40.0°C
Color Yellow
Density 1.3220g/mL
Boiling Point 213.0°C
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CH(OH)CH2OH
Refractive Index 1.4790 to 1.4810
Quantity 500 mL
Merck Index 15, 2146
Specific Gravity 1.322
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,methanol and ether,slightly soluble in toluene
Viscosity 215 mPa.s (20°C)
Formula Weight 110.54
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Chloro-1, 2-propanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.