missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1,2-propanediol 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30320959
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320959 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320959 Supplier Dr. Ehrenstorfer Supplier No. DREA11502630AL100
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Chloropropane-1,2-diol
CAS 96-24-2
Analyte or Component Names 3-Chloropropane-1,2-diol
Shipping Condition Room Temperature
Molecular Formula C3 H7 Cl O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 3-monochloropropanediol, 3-MCPD, 3-Chloro-1,2-propanediol, (RS)-3-Chloro-1,2-propanediol, (RS)-α-Chlorohydrin, (±)-2,3-Dihydroxychloropropane, (±)-3-Chloro-1,2-propanediol, 1,2-Dihydroxy-3-chloropropane, 1-Chloro-1-deoxyglycerol, 1-Chloro-2,3-dihydroxypropane, 1-Chloro-2,3-propanediol, 3-Chloro-1,2-dihydroxypropane, 3-Chloro-1,2-propylene glycol, 3-Chloropropanediol, 3-Chloropropylene glycol, 3-Monochloro-1,2-propanediol, Chlorohydrin, Chloropropanediol, Ekorod A, Epibloc, Glycerin epichlorohydrin, Glycerin monochlorohydrin, Glycerin α-monochlorhydrin, Glycerol 3-chlorohydrin, Glycerol monochlorohydrin, Glycerol α-chlorohydrin, Glycerol α-monochlorohydrin, Glyceryl chloride, Glyceryl α-chlorohydrin, U 5897, dl-α-Chlorohydrin, α-Chlorohydrin, α-Monochlorohydrin
InChI Formula InChI=1S/C3H7ClO2/c4-1-3(6)2-5/h3,5-6H,1-2H2
SMILES OCC(O)CCl
IUPAC Name 3-chloropropane-1,2-diol
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 110.5395
Quantity 1 mL
Formula Weight 110.0135 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.