missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1,2,4-triazole, 97+%, Thermo Scientific™

Product Code. 15419087
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15419087 1 g 1g
15429087 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15419087 Supplier Thermo Scientific Alfa Aesar Supplier No. H52387.03
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Chloro-1,2,4-triazole is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6818-99-1
Molecular Formula C2H2ClN3
Molecular Weight (g/mol) 103.51
MDL Number MFCD00128812
InChI Key QGOUKZPSCTVYLX-UHFFFAOYSA-N
Synonym 3-chloro-1,2,4-triazole, 3-chloro-1h-1,2,4-triazole, 3-chloro-4h-1,2,4-triazole, 1h-1,2,4-triazole, 3-chloro, 3-chloro-s-triazole, s-triazole, 3-chloro, 1h-1,2,4-triazole, 5-chloro, 3-chloro-1h-1,2,4 triazole, 3-chloro-2h-1,2,4-triazole
PubChem CID 81265
IUPAC Name 5-chloro-1H-1,2,4-triazole
SMILES ClC1=NC=NN1

Specifications

Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 103.5
Percent Purity ≥97%
Chemical Name or Material 3-Chloro-1,2,4-triazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.