missing translation for 'onlineSavingsMsg'
Learn More

(+)-3-Carene

Product Code. 16398297 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 16398297

Brand: Thermo Scientific Acros 463990250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 498-15-7 | C10H16 | 136.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-15-7
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00066417
InChI Key BQOFWKZOCNGFEC-BDAKNGLRSA-N
IUPAC Name (1S,6R)-3,7,7-trimethylbicyclo[4.1.0]hept-3-ene
SMILES CC1=CC[C@@H]2[C@H](C1)C2(C)C

Specifications

Density 0.8650 g/mL
Boiling Point 170.0°C
Flash Point 46°C
Quantity 25 mL
Specific Gravity 0.865
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in benzene, acetone, ethanol
Formula Weight 136.23
Chemical Name or Material (+)-3-Carene
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (+)-3-Carene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P331-Do NOT induce vomiting.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : (1S)-3,7,7-Trimethylbicyclo[4.1.0]hept-3-ene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.