missing translation for 'onlineSavingsMsg'
Learn More

3-Carene, 90%, stabilized

Product Code. 10140622 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10140622 1 L Glass bottle 1 L
10487212 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10140622 Supplier Thermo Scientific Acros Supplier No. 367960010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13466-78-9 | C10H16 | 136.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13466-78-9 , 138-86-3 , 127-91-3 , 99-86-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.23
MDL Number MFCD00001315
InChI Key BQOFWKZOCNGFEC-UHFFFAOYSA-N
Synonym 3-carene, carene, 3,7,7-trimethylbicyclo 4.1.0 hept-3-ene, delta-3-carene, monoterpenes, alpha-carene, bicyclo 4.1.0 hept-3-ene, 3,7,7-trimethyl, +--delta3-carene, + car-3-ene, +--3-carene
PubChem CID 26049
ChEBI CHEBI:35661
IUPAC Name 4,7,7-trimethylbicyclo[4.1.0]hept-3-ene
SMILES CC1=CCC2C(C1)C2(C)C

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8640g/mL
Boiling Point 166°C to 170°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.4720 to 1.474
Quantity 1 L
Beilstein 05, 94
Merck Index 15, 1835
Specific Gravity 0.864
Solubility Information Solubility in water: insoluble
Stabilizer 0.01% alpha-Tocopherol
Formula Weight 136.23
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 3-Carene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.