missing translation for 'onlineSavingsMsg'
Learn More

3-(Carboxypropyl)triphenylphosphonium bromide, 97%

Product Code. 10163011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Glass bottle; 25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10163011 Glass bottle; 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10163011 Supplier Thermo Scientific Acros Supplier No. 338930250
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 17857-14-6 | C22H22BrO2P | 429.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Specifications

CAS 17857-14-6
CAS Min % 96.0
CAS Max % 100.0
Melting Point 247.0°C to 249.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C22H22BrO2P
MDL Number MFCD00274196
Quantity 25 g
Synonym 3-carboxypropyltriphenylphosphonium bromide, 3-carboxypropyl triphenylphosphonium bromide, 3-carboxypropyl triphenylphosphanium bromide, carboxypropyltriphenylphosphoniumbromide, phosphonium, 3-carboxypropyl triphenyl-, bromide, 4-hydroxy-4-oxobutyl triphenylphosphonium bromide, 3-carboxypropyl triphenylphosphonium bromide 3-carboxypropyl triphenylphosphonium bromide, acmc-1bp1u, xfzdcviyhhhybq-uhfffaoysa-m
InChI Key NKVJKVMGJABKHV-UHFFFAOYSA-N
SMILES [Br-].OC(=O)CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name 3-carboxypropyl(triphenyl)phosphanium;bromide
Molecular Weight (g/mol) 429.29
PubChem CID 10717451
Formula Weight 429.29
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 3-(Carboxypropyl)triphenylphosphonium bromide, 97%
Show More Show Less
compliance-icons
Product Identifier
  • (3-Carboxypropyl)triphenylphosphonium bromide, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.