missing translation for 'onlineSavingsMsg'
Learn More

(3-Carboxypropyl)triphenylphosphonium bromide, 97%

Product Code. 11499556
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499556 500 g 500g
11479556 25 g 25g
11489556 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499556 Supplier Thermo Scientific Alfa Aesar Supplier No. A19302.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 17857-14-6 | C22H22BrO2P | 429.29 g/mol

Reactant for preparation of Piperamide analogs as histone deacetylase (HDAC) inhibitors with antitumor activity, Boron-containing benzoxaboroles as antimalarial agents, Methyl alkenyl quinolones as antimycobacterial agents, Solandelactone E via Sharpless epoxidation, Taber cyclopropanation, chemoselective reductions and lithiation-borylation-allylation sequence, Organotin compounds as antifungal agents, Oleanolic acid derivatives as inhibitors of protein tyrosine phosphatase 1B for negative regulation of insulin pathway (a promising target for treatment of diabetes and obesity), 11-oxa prostaglandin analogs with ocular hypotensive activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for preparation of Piperamide analogs as histone deacetylase (HDAC) inhibitors with antitumor activity, Boron-containing benzoxaboroles as antimalarial agents, Methyl alkenyl quinolones as antimycobacterial agents, Solandelactone E via Sharpless epoxidation, Taber cyclopropanation, chemoselective reductions and lithiation-borylation-allylation sequence, Organotin compounds as antifungal agents, Oleanolic acid derivatives as inhibitors of protein tyrosine phosphatase 1B for negative regulation of insulin pathway (a promising target for treatment of diabetes and obesity), 11-oxa prostaglandin analogs with ocular hypotensive activity.

Solubility
Soluble in water. soluble in ethanol.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17857-14-6
Molecular Formula C22H22BrO2P
Molecular Weight (g/mol) 429.29
MDL Number MFCD00274196
InChI Key NKVJKVMGJABKHV-UHFFFAOYSA-N
Synonym 3-carboxypropyltriphenylphosphonium bromide, 3-carboxypropyl triphenylphosphonium bromide, 3-carboxypropyl triphenylphosphanium bromide, carboxypropyltriphenylphosphoniumbromide, phosphonium, 3-carboxypropyl triphenyl-, bromide, 4-hydroxy-4-oxobutyl triphenylphosphonium bromide, 3-carboxypropyl triphenylphosphonium bromide 3-carboxypropyl triphenylphosphonium bromide, acmc-1bp1u, xfzdcviyhhhybq-uhfffaoysa-m
PubChem CID 10717451
IUPAC Name 3-carboxypropyl(triphenyl)phosphanium;bromide
SMILES [Br-].OC(=O)CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 244°C to 250°C
Quantity 500 g
Beilstein 4173599
Sensitivity Hygroscopic
Solubility Information Soluble in water. soluble in ethanol.
Formula Weight 429.3
Percent Purity 97%
Chemical Name or Material (3-Carboxypropyl)triphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.