missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxyphenyl isothiocyanate, 97%

Product Code. 11317706
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317706 1 g 1g
11327706 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317706 Supplier Thermo Scientific Alfa Aesar Supplier No. L11322.03
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2131-63-7 | C8H5NO2S | 179.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2131-63-7
Molecular Formula C8H5NO2S
Molecular Weight (g/mol) 179.193
MDL Number MFCD00041367
InChI Key PJRBPKOOGLKPFB-UHFFFAOYSA-N
Synonym 3-carboxyphenyl isothiocyanate, 3-carboxyphenylisothiocyanate, benzoic acid, 3-isothiocyanato, benzoic acid,3-isothiocyanato, acmc-20an7x, m-carboxyphenyl isothiocyanate, 3-isothiocyanato-benzoic acid, #
PubChem CID 75046
IUPAC Name 3-isothiocyanatobenzoic acid
SMILES C1=CC(=CC(=C1)N=C=S)C(=O)O

Specifications

Melting Point ∽165°C (decomposition)
Odor Irritating
Quantity 1 g
UN Number UN2923
Beilstein 2209003
Sensitivity Moisture sensitive
Formula Weight 179.2
Percent Purity 97%
Chemical Name or Material 3-Carboxyphenyl isothiocyanate
compliance-icons compliance-icons
Product Identifier
  • 3-Carboxyphenyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 3-Carboxyphenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.