missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxy-4-fluorophenylboronic acid, 97%, Thermo Scientific™

Product Code. 10115657 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10115657 10 g Glass bottle 10g
10737518 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10115657 Supplier Thermo Scientific Supplier No. 430220100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 872460-12-3
Molecular Formula C7H6BFO4
Molecular Weight (g/mol) 183.93
MDL Number MFCD03095129
InChI Key DGORTXQWDDXSIQ-UHFFFAOYSA-N
Synonym 3-carboxy-4-fluorophenylboronic acid, 3-carboxy-4-fluorobenzeneboronic acid, 5-dihydroxyboranyl-2-fluorobenzoic acid, 4-fluoro-3-carboxyphenylboronic acid, benzoic acid, 5-borono-2-fluoro, 3-carboxyl-4-fluorophenylboronic acid, 5-borono-2-fluoro-benzoic acid, akos brn-0540, pubchem16787, acmc-209qj6
PubChem CID 2763242
IUPAC Name 5-(dihydroxyboranyl)-2-fluorobenzoic acid
SMILES OB(O)C1=CC(C(O)=O)=C(F)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 196.0°C to 204.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 183.93
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 3-Carboxy-4-fluorophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Carboxy-4-fluorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.