missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Carboxy-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
£88.81 - £265.00
Chemical Identifiers
CAS | 1373168-89-8 |
---|---|
Molecular Formula | C13H16BFO4 |
Molecular Weight (g/mol) | 266.075 |
InChI Key | WVKOYVWFDCAHKF-UHFFFAOYSA-N |
Synonym | 2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxy-2-fluorophenylboronic acid, pinacol ester, 2-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid |
PubChem CID | 56737743 |
IUPAC Name | 2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)C(=O)O)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1373168-89-8 | |
266.075 | |
2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxy-2-fluorophenylboronic acid, pinacol ester, 2-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid | |
2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid |
C13H16BFO4 | |
WVKOYVWFDCAHKF-UHFFFAOYSA-N | |
56737743 | |
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)C(=O)O)F |
Specifications
1373168-89-8 | |
2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxy-2-fluorophenylboronic acid, pinacol ester, 2-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid | |
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)C(=O)O)F | |
266.075 | |
266.07 | |
Solid |
C13H16BFO4 | |
WVKOYVWFDCAHKF-UHFFFAOYSA-N | |
2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid | |
56737743 | |
96% | |
3-Carboxy-2-fluorobenzeneboronic acid pinacol ester |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures