missing translation for 'onlineSavingsMsg'
Learn More

3-Carbamoyl-1-methyl-d3-pyridinium Chloride, TRC

Product Code. 30484090
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30484090 10 mg 10mg
30468321 25 mg 25mg
30469436 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30484090 Supplier Toronto Research Chemicals Supplier No. TRCM32099610MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1218993-18-0
Molecular Formula C7 D3 H6 N2 O . Cl
Molecular Weight (g/mol) 175.631
Synonym Pyridinium, 3-(aminocarbonyl)-1-(methyl-d3)-, chloride (1:1) (ACI), N-Methylnicotinamide-d3 chloride, 1-Methylnicotinamide-d3 chloride, 1-(Methyl-d3)nicotinamide chloride, 1-Methylnicotinamide-D3 (methyl-D3) chloride, 3-(Aminocarbonyl)-1-(methyl-d3)pyridinium chloride, N-(Methyl-d3)nicotinamide chloride, Nicotinamide methochloride-d3, 3-Carbamoyl-1-methyl-d3-pyridinium Chloride
IUPAC Name 1-(trideuteriomethyl)pyridin-1-ium-3-carboxamide;chloride
SMILES [Cl-].[2H]C([2H])([2H])[n+]1cccc(c1)C(=O)N

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C7H8N2O.ClH/c1-9-4-2-3-6(5-9)7(8)10;/h2-5H,1H3,(H-,8,10);1H/i1D3;
Formula Weight 175.059
Chemical Name or Material 3-Carbamoyl-1-methyl-d3-pyridinium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.