missing translation for 'onlineSavingsMsg'
Learn More

3-Carbamothioylbenzeneboronic acid, 97%, Thermo Scientific™

Catalog Number 15469357
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
850568-10-4
C7H8BNO2S
181.016
MFCD06659873
ZPKLUYJBCAHWIW-UHFFFAOYSA-N
3-boronobenzothioamide, 3-carbamothioyl benzeneboronic acid, acmc-209q1a, 3-boronothiobenzamide, 3-carbamothioylphenyl boronic acid, 3-thiocarbamoylbenzeneboronic acid, boronic acid,b-3-aminothioxomethyl phenyl
44119138
(3-carbamothioylphenyl)boronic acid
B(C1=CC(=CC=C1)C(=S)N)(O)O
This item is not returnable. View return policy
850568-10-4
C7H8BNO2S
181.016
MFCD06659873
ZPKLUYJBCAHWIW-UHFFFAOYSA-N
3-boronobenzothioamide, 3-carbamothioyl benzeneboronic acid, acmc-209q1a, 3-boronothiobenzamide, 3-carbamothioylphenyl boronic acid, 3-thiocarbamoylbenzeneboronic acid, boronic acid,b-3-aminothioxomethyl phenyl
44119138
(3-carbamothioylphenyl)boronic acid
B(C1=CC(=CC=C1)C(=S)N)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 188°C to 192°C
Quantity 1 g
Formula Weight 181.02
Percent Purity 97%
Chemical Name or Material 3-Carbamothioylbenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'