missing translation for 'onlineSavingsMsg'
Learn More

(±)-3-Butyn-2-ol, 98%

Product Code. 11413854
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
250 g
Unit Size:
100g
10g
250g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413854 10 g 10g
11329509 5 g 5g
11339509 25 g 25g
11349509 100 g 100g
11423854 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11413854 Supplier Thermo Scientific Alfa Aesar Supplier No. A11881.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2028-63-9 | C4H6O | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 2028-63-9
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00004541
InChI Key GKPOMITUDGXOSB-UHFFFAOYNA-N
Synonym 3-butyn-2-ol, 3-butyne-2-ol, 3-hydroxy-1-butyne, 1-butyn-3-ol, 1-methylpropargyl alcohol, methylethynylcarbinol, 1-ethynylethanol, 1-methyl-2-propynyl alcohol, alpha-methylpropargyl alcohol, dl-3-butyn-2-ol
PubChem CID 16239
IUPAC Name but-3-yn-2-ol
SMILES CC(O)C#C

Specifikationer

Density 0.894
Boiling Point 108°C to 109°C
Flash Point 28°C (82°F)
Refractive Index 1.426
Quantity 10 g
UN Number UN2929
Beilstein 635722
Formula Weight 70.09
Percent Purity 98%
Chemical Name or Material (±)-3-Butyn-2-ol
compliance-icons compliance-icons compliance-icons
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Self-reactive substance or mixture Type G
  • Skin corrosion/irritation Category 2
Faroangivelser
  • H225-Highly flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Skyddsangivelse
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.