Learn More
3-Butyn-1-ol, 98%
Description
3-butyn-1-ol is used in the study of a palladium-catalyzed coupling with beta-tetrionic acid bromide to prepare alkynyl substituted furanones. It is also used to prepare Markovnikov addition product. Further, it is used as an intermediate of an active pharmaceutical ingredient such as fexofenadine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-butyn-1-ol is used in the study of a palladium-catalyzed coupling with beta-tetrionic acid bromide to prepare alkynyl substituted furanones. It is also used to prepare Markovnikov addition product. Further, it is used as an intermediate of an active pharmaceutical ingredient such as fexofenadine.
Solubility
Miscible with alcohols and organic solvents. Immiscible with aliphatic hydrocarbon solvents.
Notes
Incompatible with strong oxidizing agents, acids, acid anhydrides and copper.
Specifications
Specifications
| Melting Point | -63°C |
| Density | 0.927 |
| Boiling Point | 129°C to 130°C |
| Flash Point | 36°C (96°F) |
| Odor | Characteristic |
| Linear Formula | HC≡CCH2CH2OH |
| Refractive Index | 1.4405 |
| Quantity | 25 g |
| UN Number | UN1987 |
| Beilstein | 773710 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.