missing translation for 'onlineSavingsMsg'
Learn More

3-Buten-2-ol, 97%

Product Code. p-3211873 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 10107193

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 598-32-3 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-32-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00004543
InChI Key MKUWVMRNQOOSAT-UHFFFAOYNA-N
Synonym 3-buten-2-ol, 1-buten-3-ol, methyl vinylcarbinol, 3-butene-2-ol, 3-hydroxy-1-butene, propenol, 1-methyl, methyl vinyl carbinol, 1-methyl-2-propenol, 2-methyl-2-propenol, 1-methylallyl alcohol
PubChem CID 11716
IUPAC Name but-3-en-2-ol
SMILES CC(O)C=C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -100.0°C
Color Colorless
Density 0.8300g/mL
Boiling Point 96.0°C to 98.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH(OH)CH3
Refractive Index 1.4130 to 1.4150
Quantity 5 mL
Specific Gravity 0.83
Solubility Information Solubility in water: soluble
Formula Weight 72.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Buten-2-ol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Buten-2-ol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.