missing translation for 'onlineSavingsMsg'
Learn More

3-Buten-1-ol, 98+%

Product Code. 11380194
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11380194 5 g 5g
11390194 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11380194 Supplier Thermo Scientific Alfa Aesar Supplier No. L00767.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-27-0 | C4H8O | 72.107 g/mol

3-Buten-1-ol is employed in a study of the Mn-catalyzed hydrohydrazination of olefins. Also used in a study of the conversion of propargylic acetates to ethers catalyzed by ferric chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Buten-1-ol is employed in a study of the Mn-catalyzed hydrohydrazination of olefins. Also used in a study of the conversion of propargylic acetates to ethers catalyzed by ferric chloride.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-27-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00002959
InChI Key ZSPTYLOMNJNZNG-UHFFFAOYSA-N
Synonym 3-buten-1-ol, allylcarbinol, 3-butenyl alcohol, 1-buten-4-ol, vinylethyl alcohol, 3-butene-1-ol, ch2=chch2ch2oh, homoallyl alcohol, unii-3db2krm1i9, 1-hydroxy-3-butene
PubChem CID 69389
IUPAC Name but-3-en-1-ol
SMILES C=CCCO

Specifications

Density 0.841
Boiling Point 113°C to 114°C
Flash Point 32°C (89°F)
Refractive Index 1.421
Quantity 5 g
UN Number UN1987
Beilstein 1633504
Solubility Information Soluble in water.
Formula Weight 72.11
Percent Purity ≥98%
Chemical Name or Material 3-Buten-1-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.