missing translation for 'onlineSavingsMsg'
Learn More

3-Buten-1-ol, 98+%

Product Code. 10287073 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10287073 100 mL Glass bottle 100mL
10522841 25 mL Glass bottle 25mL
10503894 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10287073 Supplier Thermo Scientific Acros Supplier No. 295751000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-27-0 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-27-0
MDL Number MFCD00002959
InChI Key ZSPTYLOMNJNZNG-UHFFFAOYSA-N
Synonym 3-buten-1-ol, allylcarbinol, 3-butenyl alcohol, 1-buten-4-ol, vinylethyl alcohol, 3-butene-1-ol, ch2=chch2ch2oh, homoallyl alcohol, unii-3db2krm1i9, 1-hydroxy-3-butene
PubChem CID 69389
IUPAC Name but-3-en-1-ol
SMILES C=CCCO

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8300g/mL
Boiling Point 112.0°C to 114.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Molecular Formula C4H8O
Linear Formula H2C=CHCH2CH2OH
Refractive Index 1.4200 to 1.4220
Quantity 100 mL
Beilstein 01, 441
Specific Gravity 0.83
Solubility Information Solubility in water: soluble in water. Other solubilities: miscible with ethanol,ether,acetone
Molecular Weight (g/mol) 72.11
Formula Weight 72.11
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 3-Buten-1-ol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Buten-1-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.