missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic acid, 99%, extra pure

Product Code. 10346910 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
250mL
5 L
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10346910 250 mL Glass bottle 250mL
10336910 5 mL Glass bottle 5mL
10765581 1 L Glass bottle 1 L
10002923 5 L Plastic drum 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10346910 Supplier Thermo Scientific Acros Supplier No. 125612500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 75-75-2 | CH4O3S | 96.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 75-75-2
Molecular Formula CH4O3S
Molecular Weight (g/mol) 96.1
MDL Number MFCD00007518
InChI Key AFVFQIVMOAPDHO-UHFFFAOYSA-N
Synonym methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane
PubChem CID 6395
ChEBI CHEBI:27376
IUPAC Name methanesulfonic acid
SMILES CS(=O)(=O)O

Spécification

Melting Point 19.0°C
Density 1.4810g/mL
Boiling Point 167.0°C (10.0 mmHg)
Flash Point 189°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3SO3H
Refractive Index 1.4252 to 1.4315
Quantity 250 mL
Beilstein 04, 4
Fieser 01,666; 02,270; 04,326; 10,256; 11,321; 12,212; 13,176
Merck Index 15, 6026
Specific Gravity 1.481
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and ethanol, insoluble in hexane and methylcyclopentane, 1.50wt% benzene -0.23wt% 0-chlorotoluene (26-28°C), 0.38wt% toluene -0.47wt% ethyl disulfide (26-28°C)
Viscosity 11.6 mPa.s (25°C)
Formula Weight 96.1
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Methanesulfonic acid
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • Methanesulfonic acid
Mention d'avertissement
  • Danger
Catégorie de danger
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Mention de danger
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Conseils de prudence
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.