missing translation for 'onlineSavingsMsg'
Learn More

3-Buten-1-amine, tech. 90%

Product Code. 11378966
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378966 1 g 1g
11388966 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378966 Supplier Thermo Scientific Alfa Aesar Supplier No. L20157.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2524-49-4 | C4H9N | 71.12 g/mol

3-Buten-1-amine is an aliphatic terminal amine used for organic synthesis and in proteomics research. Its hydrochloride salt is used to improve the detection of neurodegenerative alzheimer’s disease in ELISA titrations by reducing the background due to non-specific associations of molecules and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Buten-1-amine is an aliphatic terminal amine used for organic synthesis and in proteomics research. Its hydrochloride salt is used to improve the detection of neurodegenerative alzheimer’s disease in ELISA titrations by reducing the background due to non-specific associations of molecules and proteins.

Solubility
Miscible with water.

Notes
Incompatible with strong acids, strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2524-49-4
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.12
MDL Number MFCD03425859
InChI Key ASVKKRLMJCWVQF-UHFFFAOYSA-N
Synonym 3-buten-1-amine, homoallylamine, 3-butenylamine, but-3-enylamine, acmc-1b0ze, 4-amino-1-butene, 3-buten-1-amine 9ci, n-but-3-enylamine, 3-butene-1-amine, 1-aminobut-3-ene
PubChem CID 443732
ChEBI CHEBI:31108
IUPAC Name but-3-en-1-amine
SMILES NCCC=C

Specifications

Density 0.777
Boiling Point 74°C to 77°C
Flash Point −5°C (23°F)
Odor Amine-like
Refractive Index 1.424
Quantity 1 g
UN Number UN2733
Beilstein 1735803
Sensitivity Air Sensitive
Solubility Information Miscible with water.
Formula Weight 71.12
Percent Purity 97%
Chemical Name or Material 3-Buten-1-amine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.