missing translation for 'onlineSavingsMsg'
Learn More

3-Bromotoluene, 99%, Thermo Scientific Chemicals

Catalog Number 10205850
Click to view available options
Quantity:
10 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
591-17-3
C7H7Br
171.04
MFCD00000085
WJIFKOVZNJTSGO-UHFFFAOYSA-N
3-bromotoluene, m-bromotoluene, 3-methylbromobenzene, benzene, 1-bromo-3-methyl, m-tolyl bromide, toluene, m-bromo, 5-bromotoluene, 3-bromo-1-methylbenzene, m-methylbromobenzene, 3-methyl-1-bromobenzene
11560
1-bromo-3-methylbenzene
CC1=CC=CC(Br)=C1
This item is not returnable. View return policy
591-17-3
C7H7Br
171.04
MFCD00000085
WJIFKOVZNJTSGO-UHFFFAOYSA-N
3-bromotoluene, m-bromotoluene, 3-methylbromobenzene, benzene, 1-bromo-3-methyl, m-tolyl bromide, toluene, m-bromo, 5-bromotoluene, 3-bromo-1-methylbenzene, m-methylbromobenzene, 3-methyl-1-bromobenzene
11560
1-bromo-3-methylbenzene
CC1=CC=CC(Br)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -40.0°C
Color Colorless to Yellow
Density 1.4100g/mL
Boiling Point 183.0°C to 184.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4Br
Refractive Index 1.5510 to 1.5530
Quantity 10 mL
Beilstein 05, 305
Merck Index 14, 1439
Specific Gravity 1.41
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether and benzene
Formula Weight 171.04
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Bromotoluene, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'