missing translation for 'onlineSavingsMsg'
Learn More

3-Bromotoluene, 98%

Product Code. 11481758
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481758 25 g 25g
11491758 100 g 100g
11401768 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11481758 Supplier Thermo Scientific Alfa Aesar Supplier No. A13990.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 591-17-3 | C7H7Br | 171.04 g/mol

3-Bromotoluene was used in the precise determination of bromine isotope ratio in organic compounds by MC-ICPMS ( Multicollector-Inductively Coupled Plasma Mass Spectrometer). It undergoes palladium-catalyzed cyanation reaction in the presence of K4[Fe(CN)6] as the cyanide surrogate. It also undergoes palladium-catalyzed reaction with alkynyltriarylborates to yield trisubstituted alkenylboranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromotoluene was used in the precise determination of bromine isotope ratio in organic compounds by MC-ICPMS ( Multicollector-Inductively Coupled Plasma Mass Spectrometer). It undergoes palladium-catalyzed cyanation reaction in the presence of K4[Fe(CN)6] as the cyanide surrogate. It also undergoes palladium-catalyzed reaction with alkynyltriarylborates to yield trisubstituted alkenylboranes.

Solubility
Not miscible in water. Soluble in alcohols, ether and benzene.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-17-3
Molecular Formula C7H7Br
Molecular Weight (g/mol) 171.04
MDL Number MFCD00000085
InChI Key WJIFKOVZNJTSGO-UHFFFAOYSA-N
Synonym 3-bromotoluene, m-bromotoluene, 3-methylbromobenzene, benzene, 1-bromo-3-methyl, m-tolyl bromide, toluene, m-bromo, 5-bromotoluene, 3-bromo-1-methylbenzene, m-methylbromobenzene, 3-methyl-1-bromobenzene
PubChem CID 11560
IUPAC Name 1-bromo-3-methylbenzene
SMILES CC1=CC=CC(Br)=C1

Specifications

Melting Point -40°C
Density 1.41
Boiling Point 183°C to 184°C
Flash Point 75°C (167°F)
Refractive Index 1.552
Quantity 25 g
Beilstein 1903633
Merck Index 14,1439
Solubility Information Not miscible in water. Soluble in alcohols,ether and benzene.
Formula Weight 171.04
Percent Purity 98%
Chemical Name or Material 3-Bromotoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.