missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophene, MP Biomedicals

Product Code. 11454165 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454165 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11454165 Supplier MP Biomedicals Supplier No. 0522387110
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 872-31-1
Molecular Formula C4H3BrS
MDL Number MFCD00005464
Quantity 10 g
Synonym 3-thienyl bromide, thiophene, 3-bromo, 3-bromo thiophene, beta-bromothiophene, .beta.-bromothiophene, 3-bromo-thiophene, bromothiophene 3-, 3-bromothiphene, 3-bromthiophene, pubchem5306
InChI Key XCMISAPCWHTVNG-UHFFFAOYSA-N
SMILES BrC1=CSC=C1
IUPAC Name 3-bromothiophene
Molecular Weight (g/mol) 163.03
PubChem CID 13383
Formula Weight 163.04
Chemical Name or Material 3-Bromothiophene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.