missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophene-2-sulfonyl chloride, 90%, Thermo Scientific™

Product Code. p-3540997
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10527284 250 mg Amber Glass Bottle 250mg
10757295 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10527284 Supplier Thermo Scientific Maybridge Supplier No. MO08006CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 170727-02-3
Molecular Formula C4H2BrClO2S2
Molecular Weight (g/mol) 261.532
MDL Number MFCD08059484
InChI Key OPLMNCCLQDVCTP-UHFFFAOYSA-N
Synonym 3-bromothiophene-2-sulfonylchloride, 2-thiophenesulfonylchloride, 3-bromo, 3-bromo-thiophene-2-sulfonyl chloride, 3-bromo-thiophene-2-sulphonyl chloride, 3-bromo 2-thienyl chlorosulfone, 3-bromo-2-thiophenesulfonyl chloride, 3-bromothiophene-2-sulphonyl chloride, 2-thiophenesulfonyl chloride, 3-bromo
PubChem CID 13675598
IUPAC Name 3-bromothiophene-2-sulfonyl chloride
SMILES C1=CSC(=C1Br)S(=O)(=O)Cl

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 261.55
Percent Purity 90%
Chemical Name or Material 3-Bromothiophene-2-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.