missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophene-2-carboxamide, 99%, Thermo Scientific Chemicals

Product Code. 11302907
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11302907 1 g 1g
11312907 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302907 Supplier Thermo Scientific Alfa Aesar Supplier No. L16138.03
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78031-18-2 | Molecular Formula: C5H4BrNOS | Molecular Weight: 206.06 g/mol

3-Bromothiophene-2-carboxamide is used in Stille cross-coupling conditions were used to couple the resin-bound peptide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromothiophene-2-carboxamide is used in Stille cross-coupling conditions were used to couple the resin-bound peptide.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78031-18-2
Molecular Formula C5H4BrNOS
Molecular Weight (g/mol) 206.06
MDL Number MFCD00173835
InChI Key KWXUWNXTRUZHNQ-UHFFFAOYSA-N
Synonym maybridge1_007895, 3-bromo-2-carboxamidothiophene, 2-thiophenecarboxamide,3-bromo, 3-bromo-thiophene-2-carboxamide, 3-bromo-2-thiophenecarboxamide #, buttpark 180\02-08, 3-bromo thiophene-2-carboxamide
PubChem CID 605721
IUPAC Name 3-bromothiophene-2-carboxamide
SMILES NC(=O)C1=C(Br)C=CS1

Specifications

Melting Point 106°C to 108°C
Quantity 1 g
Beilstein 4937660
Solubility Information Insoluble in water.
Formula Weight 206.06
Percent Purity 99%
Chemical Name or Material 3-Bromothiophene-2-carboxamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.