missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromothioanisole, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | 3-Bromothioanisole |
| CAS | 33733-73-2 |
| Molecular Formula | C7 H7 Br S |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C7H7BrS/c1-9-7-4-2-3-6(8)5-7/h2-5H,1H3 |
| SMILES | CSc1cccc(Br)c1 |
| IUPAC Name | 1-bromo-3-methylsulfanylbenzene |
| Molecular Weight (g/mol) | 203.1 |
| Formula Weight | 201.9452 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction