missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothioanisole, 98%, Thermo Scientific Chemicals

Product Code. 11467086
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467086 5 g 5g
11457086 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11467086 Supplier Thermo Scientific Alfa Aesar Supplier No. B25406.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33733-73-2 | Molecular Formula: C7H7BrS | Molecular Weight: 203.097 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33733-73-2
Molecular Formula C7H7BrS
Molecular Weight (g/mol) 203.097
MDL Number MFCD00041395
InChI Key NKYFJZAKUPSUSH-UHFFFAOYSA-N
Synonym 3-bromothioanisole, 1-bromo-3-methylthio benzene, 3-bromophenyl methyl sulfide, 3-bromophenyl methyl sulfane, 1-bromo-3-methylsulfanyl benzene, benzene, 1-bromo-3-methylthio, m-bromo methylthio benzene, 3-bromophenyl methyl sulphide, 1-bromo-3-methylsulfanyl-benzene, 3-bromothioanisol
PubChem CID 2735627
IUPAC Name 1-bromo-3-methylsulfanylbenzene
SMILES CSC1=CC(=CC=C1)Br

Specifications

Density 1.51
Boiling Point 124°C to 125°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.624
Quantity 5 g
Beilstein 2078598
Formula Weight 203.11
Percent Purity 98%
Chemical Name or Material 3-Bromothioanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.