missing translation for 'onlineSavingsMsg'
Learn More

3-Bromostyrene, 97%, stabilized, Thermo Scientific Chemicals

Catalog Number 11305416
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2039-86-3
C8H7Br
183.05
KQJQPCJDKBKSLV-UHFFFAOYSA-N
3-bromostyrene, 1-bromo-3-vinylbenzene, m-bromostyrene, benzene, 1-bromo-3-ethenyl, 1-bromo-3-vinyl-benzene, unii-6xe3sf241y, 3-bromo-styrene, pubchem23859, acmc-1cmdc, 1-bromo-3-vinylbenzene #
74870
1-bromo-3-ethenylbenzene
C=CC1=CC(=CC=C1)Br
This item is not returnable. View return policy
2039-86-3
C8H7Br
183.05
KQJQPCJDKBKSLV-UHFFFAOYSA-N
3-bromostyrene, 1-bromo-3-vinylbenzene, m-bromostyrene, benzene, 1-bromo-3-ethenyl, 1-bromo-3-vinyl-benzene, unii-6xe3sf241y, 3-bromo-styrene, pubchem23859, acmc-1cmdc, 1-bromo-3-vinylbenzene #
74870
1-bromo-3-ethenylbenzene
C=CC1=CC(=CC=C1)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4060g/mL
Boiling Point 74.0°C to 75.0°C (3.0 mmHg)
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHC6H4Br
Refractive Index 1.5890 to 1.5910
Quantity 1 g
Specific Gravity 1.406
Stabilizer 0.05% 3, 5-di-tert.-butylcatechol
Formula Weight 183.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromostyrene, Stabilized
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'