missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopyruvic acid hydrate, 98%

Product Code. 10328940 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10328940 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10328940 Supplier Thermo Scientific Acros Supplier No. 325690250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 206860-50-6 | C3H3BrO3 | 166.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 206860-50-6, 1113-59-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 79°C to 82°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C3H3BrO3
MDL Number MFCD00149107
Quantity 25 g
Synonym 3-bromopyruvic acid hydrate, 3-bromo-2-oxopropanoic acid hydrate, bromopyruvic acid hydrate, 3-bromo-2-oxopropanoic acid, hydrate, 3-bromo-2-oxopropanoic acid-water 1/1, propanoic acid, 3-bromo-2-oxo-, monohydrate 9ci
Solubility Information Solubility in water: soluble
InChI Key PRRZDZJYSJLDBS-UHFFFAOYSA-N
SMILES OC(=O)C(=O)CBr
IUPAC Name 3-bromo-2-oxopropanoic acid;hydrate
Molecular Weight (g/mol) 166.96
PubChem CID 44118703
Formula Weight 166.96
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Bromopyruvic acid hydrate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Bromopyruvic acid hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.