missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopyridine, 97%, Maybridge

Product Code. 10193733
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10193733 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10193733 Supplier Thermo Scientific Maybridge Supplier No. CC04110DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 626-55-1
CAS Min % 96.5
CAS Max % 100.0
Density 1.6400g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C5H4BrN
Linear Formula (C5H4N)Br
MDL Number MFCD00006373
Quantity 1 g
Synonym 3-pyridyl bromide, pyridine, 3-bromo, 3-bromo pyridine, 3-bromo-pyridine, 5-bromopyridine, unii-xmn8h2xe9h, 5-bromo pyridine, xmn8h2xe9h, 3-pyridylbromide, 3-bromopryidine
InChI Key NYPYPOZNGOXYSU-UHFFFAOYSA-N
SMILES BrC1=CC=CN=C1
IUPAC Name 3-bromopyridine
Molecular Weight (g/mol) 158.00
PubChem CID 12286
ChEBI CHEBI:51575
Formula Weight 158
Percent Purity 97%
Chemical Name or Material 3-Bromopyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.