missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromopyrazole, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
CAS | 14521-80-3 |
Melting Point | 60.0°C to 65.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass Bottle |
Molecular Formula | C3H3BrN2 |
Quantity | 1 g |
Synonym | 3-bromo-1h-pyrazole, 3-bromopyrazole, 1h-pyrazole, 3-bromo, 3-bromo-pyrazole, bromopyrazole, 5-bromopyrazole, 3-bromo-2h-pyrazole, zlchem 53, pubchem22992 |
InChI Key | XHZWFUVEKDDQPF-UHFFFAOYSA-N |
SMILES | C1=C(NN=C1)Br |
Show More |
Safety and Handling
Product Identifier
- 3-Bromopyrazole
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'