missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromopropionyl chloride, tech. 90%

Product Code. 11471870
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471870 100 g 100g
11461870 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11471870 Supplier Alfa Aesar™ Supplier No. A10846.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
3-Bromopropionyl chloride acts as an acrylate synthon by reaction with 2-oxazolidinone, followed by base-promoted elimination to form a Diels-Alder dienophile. Brominated poly (ethylene glycol) methacrylate (v-PEG-Br) which was synthesized by the reaction of poly (ethyleneglycol) methacrylate, v-PEG-OH with Mn = 526 Dalton and 3-bromopropionyl chloride. Used in the reduction of organic compounds by mixed hydrides. Used as reagent in Chroman and tetrahydroquinoline ureas as potent TRPV1 antagonists. Reaction of veratrole with 3- bromopropionyl chloride led to the formation of 15 in 82% yield, which on treatment with the Tebbe reagent13 led to the formation of styryl bromide 16 in 93% yield. Used in the preparation of ome new mono- and di-nitrogen-containing monobenzo-crowns. 3-Bromopropionyl chloride was used in the preparation of 1-chloro-4-bromo-2-butanone.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15486-96-1
Molecular Formula C3H4BrClO
Molecular Weight (g/mol) 171.42
MDL Number MFCD00000746
InChI Key IHBVNSPHKMCPST-UHFFFAOYSA-N
Synonym 3-bromopropionyl chloride, propanoyl chloride, 3-bromo, 3-bromopropionylchloride, sgqdjlpqbrmuh@, 3-bromoproionylchloride, 3-bromopropanyl chloride, 3-bromo propionylchloride, 3-bromopropionic chloride, 3-bromopropionoyl chloride, acmc-1bwl2
PubChem CID 84944
IUPAC Name 3-bromopropanoyl chloride
SMILES ClC(=O)CCBr

Specifications

Boiling Point 54°C to 56°C (14 mmHg)
Flash Point 79°C (174°F)
Quantity 100 g
UN Number UN3265
Beilstein 605453
Formula Weight 171.43
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-Bromopropionyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.