missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropionitrile, TRC

Product Code. 30343254
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 30343254

Brand: Toronto Research Chemicals TRCB67889050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Bromopropionitrile
CAS 2417-90-5
Molecular Formula C3H4BrN
Quantity 50 g
Synonym 3-Bromopropanenitrile, 1-Bromo-2-cyanoethane, 2-Bromoethyl Cyanide, 2-Cyanoethyl Bromide, 3-Bromopropionitrile, NSC 8021, beta-Bromopropionitrile
InChI Formula InChI=1S/C3H4BrN/c4-2-1-3-5/h1-2H2
SMILES BrCCC#N
IUPAC Name 3-bromopropanenitrile
Molecular Weight (g/mol) 133.97
Formula Weight 132.9527
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.