missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropionitrile, TRC

Product Code. 30347904
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30347904 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347904 Supplier Toronto Research Chemicals Supplier No. TRCB67889010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material 3-Bromopropionitrile
CAS 2417-90-5
Molecular Formula C3H4BrN
Quantity 10 g
Synonym 3-Bromopropanenitrile, 1-Bromo-2-cyanoethane, 2-Bromoethyl Cyanide, 2-Cyanoethyl Bromide, 3-Bromopropionitrile, NSC 8021, beta-Bromopropionitrile
InChI Formula InChI=1S/C3H4BrN/c4-2-1-3-5/h1-2H2
SMILES BrCCC#N
IUPAC Name 3-bromopropanenitrile
Molecular Weight (g/mol) 133.97
Formula Weight 132.9527
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.