missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropionic acid, 97%

Product Code. 11389509
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389509 25 g 25g
11484633 100 g 100g
11494633 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11389509 Supplier Thermo Scientific Alfa Aesar Supplier No. A11573.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 590-92-1 | C3H5BrO2 | 152.98 g/mol

3-Bromopropionic acid used as quaternization agent in amperometric biosensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromopropionic acid used as quaternization agent in amperometric biosensors.

Solubility
Soluble in water, alcohol, ether, chloroform, benzene

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-92-1
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00002763
InChI Key DHXNZYCXMFBMHE-UHFFFAOYSA-N
Synonym 3-bromopropionic acid, propanoic acid, 3-bromo, beta-bromopropionic acid, propionic acid, 3-bromo, 2-carboxyethyl bromide, beta-bromopropanoic acid, unii-wfz7csr69r, beta-propionic acid, 3-bromo-propionic acid, wfz7csr69r
PubChem CID 11553
IUPAC Name 3-bromopropanoic acid
SMILES OC(=O)CCBr

Specifications

Melting Point 57°C to 63°C
Density 1.48
Boiling Point 140°C to 142°C (45 mmHg)
Flash Point 65°C (149°F)
Quantity 25 g
UN Number UN3261
Beilstein 1071333
Merck Index 14,1433
Solubility Information Soluble in water, alcohol, ether, chloroform, benzene
Formula Weight 152.98
Percent Purity 97%
Chemical Name or Material 3-Bromopropionic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.