missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenylacetic acid, 98+%

Product Code. 11485626
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485626 25 g 25g
11475626 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485626 Supplier Thermo Scientific Alfa Aesar Supplier No. B20758.14
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1878-67-7 | C8H7BrO2 | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-67-7
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00004330
InChI Key KYNNBXCGXUOREX-UHFFFAOYSA-N
Synonym 3-bromophenylacetic acid, 2-3-bromophenyl acetic acid, 3-bromophenyl acetic acid, m-bromophenylacetic acid, benzeneacetic acid, 3-bromo, 3-bromophenylaceticacid, 3-bromo-phenylacetic acid, 3-bromphenylacetic acid, pubchem4244
PubChem CID 74653
IUPAC Name 2-(3-bromophenyl)acetic acid
SMILES OC(=O)CC1=CC=CC(Br)=C1

Specifications

Melting Point 100°C to 102°C
Quantity 25 g
Beilstein 2045104
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 3-Bromophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.