missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenyl isocyanate, 98%

Product Code. 11368725
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368725 5 g 5g
11358725 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368725 Supplier Thermo Scientific Alfa Aesar Supplier No. L12292.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 23138-55-8 | C7H4BrNO | 198.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23138-55-8
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.02
MDL Number MFCD00002014
InChI Key VQVBCZQTXSHJGF-UHFFFAOYSA-N
Synonym 3-bromophenyl isocyanate, 3-bromophenylisocyanate, benzene, 1-bromo-3-isocyanato, isocyanic acid 3-bromophenyl ester, 3-bromobenzenisocyanate, m-bromophenylisocyanate, 3-bromophenyliso-cyanate, 3-bromo phenyl isocyanate, 3-bromo-phenyl isocyanate, acmc-209g1f
PubChem CID 141001
IUPAC Name 1-bromo-3-isocyanatobenzene
SMILES BrC1=CC=CC(=C1)N=C=O

Specifications

Density 1.586
Boiling Point 220°C
Flash Point 108°C (226°F)
Odor Pungent
Refractive Index 1.5855
Quantity 5 g
UN Number UN2206
Beilstein 2802537
Sensitivity Moisture sensitive
Formula Weight 198.03
Percent Purity 98%
Chemical Name or Material 3-Bromophenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.