missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenetole, 98%

Product Code. 11479387
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479387 50 g 50g
11469387 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479387 Supplier Thermo Scientific Alfa Aesar Supplier No. B21418.18
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2655-84-7 | C8H9BrO | 201.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2655-84-7
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00274287
InChI Key LQBMPJSTUHWGDE-UHFFFAOYSA-N
Synonym 3-bromophenetole, benzene, 1-bromo-3-ethoxy, m-bromophenetole, 3-bromophenyl ethyl ether, 1-bromo-3-ethoxy-benzene, 3-bromoethoxybenzene, 3-bromphenyl ethyl ether, phenetole, m-bromo, m-bromphenetol, 3-ethoxybromobenzene
PubChem CID 75867
IUPAC Name 1-bromo-3-ethoxybenzene
SMILES CCOC1=CC=CC(Br)=C1

Specifications

Density 1.41
Boiling Point 231°C
Flash Point 83°C (182°F)
Refractive Index 1.548
Quantity 50 g
Formula Weight 201.06
Percent Purity 98%
Chemical Name or Material 3-Bromophenetole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.