missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenethyl alcohol, 97%

Product Code. 10591282 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10591282 1 g Glass bottle 1g
10427972 5 g Glass bottle 5g
10388983 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10591282 Supplier Thermo Scientific Acros Supplier No. 350560010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28229-69-8 | C8H9BrO | 201.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28229-69-8
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00191857
InChI Key PTTFLKHCSZSFOL-UHFFFAOYSA-N
Synonym 3-bromophenethyl alcohol, 2-3-bromophenyl ethanol, 2-3-bromophenyl ethan-1-ol, benzeneethanol, 3-bromo, 3-bromophenethylalcohol, 3-bromobenzeneethanol, 3-2-hydroxyethyl bromobenzene, 2-3-bromophenyl ethyl alcohol, pubchem14821, 3-bromo-phenethyl-alcohol
PubChem CID 2734090
IUPAC Name 2-(3-bromophenyl)ethanol
SMILES OCCC1=CC=CC(Br)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.4780g/mL
Boiling Point 107°C to 110°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5730 to 1.575
Quantity 1 g
Specific Gravity 1.478
Formula Weight 201.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromophenethyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromophenethylalcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.