missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromophenacyl Bromide, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 3-Bromophenacyl Bromide |
| CAS | 18523-22-3 |
| Molecular Formula | C8H6Br2O |
| Quantity | 250 mg |
| Synonym | 2-Bromo-1-(3-bromophenyl)ethanone, 2,3'-Dibromoacetophenone |
| InChI Formula | InChI=1S/C8H6Br2O/c9-5-8(11)6-2-1-3-7(10)4-6/h1-4H,5H2 |
| SMILES | BrCC(C1=CC(Br)=CC=C1)=O |
| IUPAC Name | 2-bromo-1-(3-bromophenyl)ethan-1-one |
| Molecular Weight (g/mol) | 277.94 |
| Formula Weight | 275.88 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction