missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)pyridine hydrobromide, 97%

Product Code. p-3770871 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10306374 1 g 1g
10420744 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10306374 Supplier Thermo Scientific Acros Supplier No. 397140010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4916-55-6 | C6H6BrN·HBr | 252.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4916-55-6
Molecular Formula C6H6BrN·HBr
Molecular Weight (g/mol) 252.93
MDL Number MFCD01321314
InChI Key FNHPUOJKUXFUKN-UHFFFAOYSA-N
Synonym 3-bromomethyl pyridine hydrobromide, 3-picolyl bromide hydrobromide, 3-bromomethylpyridine hydrobromide, 3-bromomethyl-pyridine hydrobromide, pyridine, 3-bromomethyl-, hydrobromide, pubchem17664, acmc-209kf1, 3-brommethyl pyridine-hbr, 3-bromomethyl pyridine, bromide, 3-bromomethyl pyridinehydrobromide
PubChem CID 12707037
IUPAC Name 3-(bromomethyl)pyridine;hydrobromide
SMILES C1=CC(=CN=C1)CBr.Br

Specifications

Melting Point 150.0°C to 155.0°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 252.93
Percent Purity 97%
Chemical Name or Material 3-(Bromomethyl)pyridine hydrobromide
compliance-icons compliance-icons
Product Identifier
  • 3-(Bromomethyl)pyridine hydrobromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.