missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoimidazo[1,2-a]pyridine-7-carboxylic acid, 95%

Product Code. 15469025
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15469025 250 mg 250mg
15459025 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469025 Supplier Thermo Scientific Alfa Aesar Supplier No. H35563.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1315360-75-8 | C8H5BrN2O2 | 241.044 g/mol

3-Bromoimidazo[1,2-a]pyridine-7-carboxylic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromoimidazo[1,2-a]pyridine-7-carboxylic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1315360-75-8
Molecular Formula C8H5BrN2O2
Molecular Weight (g/mol) 241.044
MDL Number MFCD21602377
InChI Key NBXDWHKFUACMKB-UHFFFAOYSA-N
Synonym 3-bromoimidazo 1,2-a pyridine-7-carboxylic acid, 3-bromo-7-carboxyimidazo 1,2-a pyridine, imidazo 1,2-a pyridine-7-carboxylic acid,3-bromo
PubChem CID 66222606
IUPAC Name 3-bromoimidazo[1,2-a]pyridine-7-carboxylic acid
SMILES C1=CN2C(=NC=C2Br)C=C1C(=O)O

Specifications

Melting Point 282°C to 285°C
Quantity 250 mg
Formula Weight 241.05
Percent Purity 95%
Chemical Name or Material 3-Bromoimidazo[1,2-a]pyridine-7-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.