missing translation for 'onlineSavingsMsg'
Learn More

3-Bromofuro[3,2-b]pyridine, 96%

Product Code. p-7032257
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15406507 250 mg 250mg
15486497 1 g 1g
15496497 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15406507 Supplier Thermo Scientific Alfa Aesar Supplier No. H33652.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 92404-63-2 | C7H4BrNO | 198.019 g/mol

3-Bromofuro[3,2-b]pyridine is used as a starting material for synthesis of polyheterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromofuro[3,2-b]pyridine is used as a starting material for synthesis of polyheterocycles.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92404-63-2
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.019
MDL Number MFCD19687763
InChI Key KHMAAFDMZSYVEF-UHFFFAOYSA-N
Synonym 3-bromofuro 3,2-b pyridine, furo 3,2-b pyridine, 3-bromo, acmc-20lvu1
PubChem CID 641477
IUPAC Name 3-bromofuro[3,2-b]pyridine
SMILES C1=CC2=C(C(=CO2)Br)N=C1

Specifications

Melting Point 51°C to 53°C
Quantity 250 mg
Formula Weight 198.02
Percent Purity 96%
Chemical Name or Material 3-Bromofuro[3,2-b]pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.