missing translation for 'onlineSavingsMsg'
Learn More

3-Bromofuran, 97%, Thermo Scientific™

Product Code. 10217232
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10217232 25 g 25g
10661046 5 g 5g
10501773 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10217232 Supplier Thermo Scientific Maybridge Supplier No. CC02110EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 22037-28-1 , 7440-50-8 , 110-00-9 , 144-55-8
Molecular Formula C4H3BrO
Molecular Weight (g/mol) 146.97
MDL Number MFCD00005347
InChI Key LXWLEQZDXOQZGW-UHFFFAOYSA-N
Synonym furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz
PubChem CID 89164
IUPAC Name 3-bromofuran
SMILES BrC1=COC=C1

Specifications

CAS Min % 94.5, 0.05, 0.45, 0.35
CAS Max % 98.5, 0.15, 0.55, 0.45
Density 1.635g/mL
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 25 g
Formula Weight 146.97
Percent Purity 95%
Chemical Name or Material 3-Bromofuran, stabilized with Cu(0.1%), Furan (0.5%) & NaHCO3 (0.4%)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.