missing translation for 'onlineSavingsMsg'
Learn More

3-Bromofluorobenzene, 99%

Product Code. 10054053 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10054053 500 mL 500mL
10427480 25 mL 25mL
10794591 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10054053 Supplier Thermo Scientific Acros Supplier No. 107005000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1073-06-9 | C6H4BrF | 175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073-06-9
Molecular Formula C6H4BrF
Molecular Weight (g/mol) 175
MDL Number MFCD00000326
InChI Key QDFKKJYEIFBEFC-UHFFFAOYSA-N
Synonym 3-bromofluorobenzene, m-bromofluorobenzene, m-fluorobromobenzene, benzene, 1-bromo-3-fluoro, 3-bromfluorbenzen, 3-fluorobromobenzene, m-fluorophenyl bromide, 1-fluoro-3-bromobenzene, 3-fluoro bromobenzene, benzene, 1-fluoro-3-bromo
PubChem CID 14082
IUPAC Name 1-bromo-3-fluorobenzene
SMILES C1=CC(=CC(=C1)Br)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5670g/mL
Boiling Point 150°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula BrC6H4F
Refractive Index 1.5260 to 1.528
Quantity 500 mL
Specific Gravity 1.567
Solubility Information Solubility in water: insoluble in water. Other solubilities: organic soluble/miscible with most solvents
Formula Weight 175
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Bromofluorobenzene, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Bromofluorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.