missing translation for 'onlineSavingsMsg'
Learn More

3-Bromocyclohexene, 95%

Product Code. 11454183
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454183 100 g 100g
11434183 5 g 5g
11444183 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454183 Supplier Thermo Scientific Alfa Aesar Supplier No. A10370.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1521-51-3 | C6H9Br | 161.042 g/mol

In peptide synthesis, can be used to protect the hydroxyl group of serine and threonine as the cyclohexyl (Chx) ether, by reaction with the sodium alkoxide (generated with NaH), followed by hydrogenation. The group is stable to TFA and 20% piperidine in DMF, but can be cleaved with TfOH in TFA, in the presence of thioanisole as a cation scavenger. It is also used in the synthesis of N-tert-butoxycarbonyl-O-cyclohexyl-L-tyrosine.1 It was also used in the synthesis of enantiopure cyclohexitols such as muco-quercitol, D-chiro-inocitol and allo-inocitol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In peptide synthesis, can be used to protect the hydroxyl group of serine and threonine as the cyclohexyl (Chx) ether, by reaction with the sodium alkoxide (generated with NaH), followed by hydrogenation. The group is stable to TFA and 20% piperidine in DMF, but can be cleaved with TfOH in TFA, in the presence of thioanisole as a cation scavenger. It is also used in the synthesis of N-tert-butoxycarbonyl-O-cyclohexyl-L-tyrosine.1 It was also used in the synthesis of enantiopure cyclohexitols such as muco-quercitol, D-chiro-inocitol and allo-inocitol.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1521-51-3
Molecular Formula C6H9Br
Molecular Weight (g/mol) 161.042
MDL Number MFCD00013775
InChI Key AJKDUJRRWLQXHM-UHFFFAOYSA-N
Synonym 3-bromocyclohex-1-ene, cyclohexene, 3-bromo, 1-bromo-2-cyclohexene, 3-bromo-1-cyclohexene, 1,2,3,4-tetrahydrobromobenzene, 3-bromo-cyclohexene, 2-cyclohexenyl bromide, 2-cyclohexen-1-yl bromide, 3-bromo cyclohexene, 3-bromanylcyclohexene
PubChem CID 137057
IUPAC Name 3-bromocyclohexene
SMILES C1CC=CC(C1)Br

Specifications

Density 1.4
Boiling Point 64°C to 65°C (15 mmHg)
Flash Point 54°C (129°F)
Refractive Index 1.531
Quantity 100 g
UN Number UN1993
Beilstein 635953
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 161.05
Percent Purity 95%
Chemical Name or Material 3-Bromocyclohexene, Stabilized with 300 to 1000ppm Propylene oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.