missing translation for 'onlineSavingsMsg'
Learn More

3-Bromocatechol, 95%

Product Code. p-7029461
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15413567

Brand: Thermo Scientific Alfa Aesar H26925.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14381-51-2 | C6H5BrO2 | 189.008 g/mol

3-Bromocatechol, is used as a drug and agrochemicals intermediate, Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromocatechol, is used as a drug and agrochemicals intermediate, Pharmaceutical intermediate.

Solubility
Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14381-51-2
Molecular Formula C6H5BrO2
Molecular Weight (g/mol) 189.008
MDL Number MFCD00869768
InChI Key JPBDMIWPTFDFEU-UHFFFAOYSA-N
Synonym 3-bromocatechol, 3-bromopyrocatechol, 1-bromo-2,3-dihydroxybenzene, 1,2-benzenediol, 3-bromo, benzenediol, bromo, 3-bromo-1,2-benzenediol, 2,3-dihydroxy-bromobenzene, bromocatechol, 3-bromoatechol, pubchem19085
PubChem CID 26659
IUPAC Name 3-bromobenzene-1,2-diol
SMILES C1=CC(=C(C(=C1)Br)O)O

Specifications

Melting Point 39°C to 42°C
Quantity 250 mg
Solubility Information Soluble in organic solvents.
Formula Weight 189.01
Percent Purity 95%
Chemical Name or Material 3-Bromocatechol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.