missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromobenzylamine, 95%, Thermo Scientific™
£18.90 - £164.30
Chemical Identifiers
CAS | 10269-01-9 |
---|---|
Molecular Formula | C7H8BrN |
Molecular Weight (g/mol) | 186.05 |
MDL Number | MFCD01026119 |
InChI Key | SUYJXERPRICYRX-UHFFFAOYSA-N |
Synonym | 3-bromobenzylamine, 3-bromophenyl methanamine, benzenemethanamine, 3-bromo, 3-bromophenyl methylamine, m-bromobenzylamine, 1-3-bromophenyl methanamine, 3-bromo-benzylamine, 3-bromobenzenemethanamine, 3-bromobenzyl amine, pubchem21109 |
PubChem CID | 457587 |
IUPAC Name | (3-bromophenyl)methanamine |
SMILES | C1=CC(=CC(=C1)Br)CN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10078269
|
Thermo Scientific Acros
428960010 |
1g | Glass bottle |
£18.90
1g |
Estimated Shipment: 14-07-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10627970
|
Thermo Scientific Acros
428960050 |
5g | Glass bottle |
£39.10
5g |
Estimated Shipment: 14-07-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10159679
|
Thermo Scientific Acros
428960250 |
25g | Glass bottle |
£164.30
25g |
Estimated Shipment: 14-07-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
10269-01-9 | |
186.05 | |
SUYJXERPRICYRX-UHFFFAOYSA-N | |
457587 | |
C1=CC(=CC(=C1)Br)CN |
C7H8BrN | |
MFCD01026119 | |
3-bromobenzylamine, 3-bromophenyl methanamine, benzenemethanamine, 3-bromo, 3-bromophenyl methylamine, m-bromobenzylamine, 1-3-bromophenyl methanamine, 3-bromo-benzylamine, 3-bromobenzenemethanamine, 3-bromobenzyl amine, pubchem21109 | |
(3-bromophenyl)methanamine |
Specifications
3-Bromobenzylamine | |
94.0 | |
Colorless to Yellow | |
Authentic | |
BrC6H4CH2NH2 | |
Glass bottle | |
3-bromobenzylamine, 3-bromophenyl methanamine, benzenemethanamine, 3-bromo, 3-bromophenyl methylamine, m-bromobenzylamine, 1-3-bromophenyl methanamine, 3-bromo-benzylamine, 3-bromobenzenemethanamine, 3-bromobenzyl amine, pubchem21109 | |
SUYJXERPRICYRX-UHFFFAOYSA-N | |
(3-bromophenyl)methanamine | |
457587 | |
Liquid |
10269-01-9 | |
100.0 | |
94% min. (GC) | |
C7H8BrN | |
MFCD01026119 | |
1.5850 to 1.5900 (20°C, 589nm) | |
1g | |
C1=CC(=CC(=C1)Br)CN | |
186.05 | |
186.05 | |
95% |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 3-Bromobenzylamine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only